1-Bromo-3-n-octylbenzene ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
B4817-1G
B4817-5G
TCB4817-1GEA
84
USD
TCB4817-1G
TCB4817-5G
1-Bromo-3-n-octylbenzene ≥97.0% (by GC)
1-Bromo-3-n-octylbenzene
Formula:
C₁₄H₂₁Br MW: 269.23 g/mol |
CAS Number:
663949-34-6 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...