o-Tolyl trifluoromethanesulfonate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Trifluoromethanesulphonic acid o-Tolyl ester, o-tolyl Trifluoromethanesulphonate, o-Tolyl triflate

T3357-25G T3357-5G
TCT3357-25GEA 351 USD
TCT3357-25G TCT3357-5G
o-Tolyl trifluoromethanesulfonate ≥98.0% (by GC)
o-Tolyl trifluoromethanesulfonate
Formula: C₈H₇F₃O₃S
MW: 240.20 g/mol
MDL Number: MFCD29089375
CAS Number: 66107-34-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR