6-Methoxyquinoline N-Oxide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 6-Methoxyquinoline 1-oxide

M2999-1G M2999-5G
TCM2999-1GEA 74 USD
TCM2999-1G TCM2999-5G
6-Methoxyquinoline N-Oxide ≥98.0% (by GC)
6-Methoxyquinoline N-Oxide
Formula: C₁₀H₉NO₂
MDL Number: MFCD00009771
CAS Number: 6563-13-9

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 108.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR