1-Allyl-3-methylimidazolium bis(trifluoromethanesulfonyl)imide ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
1-Allyl-3-methylimidazolium bis((trifluoromethyl)sulphonyl)imide
A3210-25G
A3210-5G
TCA3210-25GEA
288
USD
TCA3210-25G
TCA3210-5G
1-Allyl-3-methylimidazolium bis(trifluoromethanesulfonyl)imide ≥98.0% (by HPLC)
1-Allyl-3-methylimidazolium bis(trifluoromethanesulfonyl)imide
Formula:
C₉H₁₁F₆N₃O₄S₂ MW: 403.31 g/mol |
MDL Number:
MFCD11109343 CAS Number: 655249-87-9 |
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...