2,7-Dibromo-9-hexylcarbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

D5055-1G D5055-5G
76219-816EA 58 USD
76219-816 76219-818
2,7-Dibromo-9-hexylcarbazole ≥98.0% (by GC)
2,7-Dibromo-9-hexylcarbazole
Formula: C₁₈H₁₉Br₂N
MDL Number: MFCD29089359
CAS Number: 654676-12-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR