Tris(4-biphenylyl)amine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Tri([1,1'-biphenyl]-4-yl)amine

T3327-1G T3327-5G
TCT3327-1GEA 133 USD
TCT3327-1G TCT3327-5G
Tris(4-biphenylyl)amine ≥98.0% (by HPLC)
Tris(4-biphenylyl)amine
Formula: C₃₆H₂₇N
MW: 473.62 g/mol
MDL Number: MFCD30470498
CAS Number: 6543-20-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR