4'-Methyl-4-pentylbiphenyl ≥98.0% (by GC)

Supplier: TCI America

M2885-1G M2885-5G
TCM2885-1GEA 71 USD
TCM2885-1G TCM2885-5G
4'-Methyl-4-pentylbiphenyl ≥98.0% (by GC)
4'-Methyl-4-pentylbiphenyl
Formula: C₁₈H₂₂
MDL Number: MFCD11053476
CAS Number: 64835-63-8
EINECS: 265-248-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR