3-(1,3-Benzodioxol-5-yl)-1-phenyl-2-propen-1-one ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3,4-(Methylenedioxy)chalcone

B4983-1G B4983-5G
TCB4983-1GEA 36 USD
TCB4983-1G TCB4983-5G
3-(1,3-Benzodioxol-5-yl)-1-phenyl-2-propen-1-one ≥98.0% (by GC)
3-(1,3-Benzodioxol-5-yl)-1-phenyl-2-propen-1-one
Formula: C₁₆H₁₂O₃
MW: 252.27 g/mol
MDL Number: MFCD00022990
CAS Number: 644-34-8

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR