N,N'-Bis(4-methoxyphenyl)-3,4,9,10-perylenetetracarboxylic diimide ≥97.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Pigment Red 190

B2892-1G B2892-5G
TCB2892-1GEA 88 USD
TCB2892-1G TCB2892-5G
N,N'-Bis(4-methoxyphenyl)-3,4,9,10-perylenetetracarboxylic diimide ≥97.0% (by total nitrogen basis)
N,N'-Bis(4-methoxyphenyl)-3,4,9,10-perylenetetracarboxylic diimide
Formula: C₃₈H₂₂N₂O₆
MW: 602.60 g/mol
MDL Number: MFCD00071978
CAS Number: 6424-77-7
EINECS: 229-187-9

Order Now

Specification Test Results

Appearance Yellow to Amber to Dark red powder to crystal
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR