4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 4-Methyl-2-thiopheneboronic acid Pinacol ester, 4-Methylthiophene-2-boronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-(4-methylthiophen-2-yl)-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(4-methyl-2-thienyl)-1,3,2-dioxaborolane

M2647-1G M2647-5G
TCM2647-1GEA 41 USD
TCM2647-1G TCM2647-5G
4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥97.0% (by GC)
4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
Formula: C₁₁H₁₇BO₂S
MW: 224.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11045430
CAS Number: 635305-48-5

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR