Dimethyl Dipropargylmalonate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Dipropargylmalonic acid Dimethyl ester, Dimethyl 2,2-di(prop-2-yn-1-yl)malonate

D4963-1G D4963-5G
TCD4963-1GEA 187 USD
TCD4963-1G TCD4963-5G
Dimethyl Dipropargylmalonate ≥98.0% (by GC)
Dimethyl Dipropargylmalonate
Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
MDL Number: MFCD00455571
CAS Number: 63104-44-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR