9-(4-Biphenylyl)carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B5128-1G B5128-5G
TCB5128-1GEA 34 USD
TCB5128-1G TCB5128-5G
9-(4-Biphenylyl)carbazole ≥98.0% (by GC)
9-(4-Biphenylyl)carbazole
Formula: C₂₄H₁₇N
MW: 319.41 g/mol
MDL Number: MFCD28147704
CAS Number: 6299-16-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 225.0 to 229.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR