3-Bromo-9-n-octyl-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B5610-1G B5610-5G
TCB5610-1GEA 152 USD
TCB5610-1G TCB5610-5G
3-Bromo-9-n-octyl-9H-carbazole ≥98.0% (by GC)
3-Bromo-9-n-octyl-9H-carbazole
Formula: C₂₀H₂₄BrN
Boiling Pt: 135°C
MDL Number: MFCD31618112
CAS Number: 628337-00-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR