sec-Butyl Sulfide ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Di-sec-Butyl Sufide, 3,5-Dimethyl-4-thiaheptane, Di-sec-butylsulphide, 2-(sec-Butylthio)butane

B6136-25ML B6136-5ML
TCB6136-25MLEA 165 USD
TCB6136-25ML TCB6136-5ML
sec-Butyl Sulfide ≥98.0% (by GC)
sec-Butyl Sulfide
Formula: C₈H₁₈S
MW: 146.29 g/mol
Boiling Pt: 165°C
Flash Pt: 39°C
MDL Number: MFCD00009332
CAS Number: 626-26-6
EINECS: 210-938-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR