Gastrodin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-(Hydroxymethyl)phenyl beta-D-Glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triol

G0468-1G G0468-200MG
TCG0468-1GEA 441.5 USD
TCG0468-1G TCG0468-200MG
Gastrodin ≥98.0% (by HPLC)
Gastrodin
Formula: C₁₃H₁₈O₇
MW: 286.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00272169
CAS Number: 62499-27-8
Merck Index: 14,04375

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 153.0 to 157.0 deg-C
Specific rotation [a]20/D -67.0 to -71.0 deg (C=1,H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR