Ethylenediamine dihydrobromide ≥97.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

E1221-1G E1221-5G
TCE1221-1GEA 73 USD
TCE1221-1G TCE1221-5G
Ethylenediamine dihydrobromide ≥97.0% (by total nitrogen and titration analysis)
Ethylenediamine dihydrobromide
Formula: C₂H₈N₂·₂HBr
Melting Pt: 350°C
CAS Number: 624-59-9
EINECS: 210-853-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR