2,7-Dibromophenanthrene ≥98.0% (by GC)
Supplier: TCI America
D5050-1G
D5050-200MG
76219-810EA
503
USD
76219-810
76219-812
2,7-Dibromophenanthrene ≥98.0% (by GC)
2,7-Dibromophenanthrene
Formula:
C₁₄H₈Br₂ MW: 336.03 g/mol |
MDL Number:
MFCD09909861 CAS Number: 62325-30-8 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 203.0 to 207.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...