4-Bromobenz[a]anthracene ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 4-Bromotetraphene

B5450-1G B5450-200MG
TCB5450-1GEA 75 USD
TCB5450-1G TCB5450-200MG
4-Bromobenz[a]anthracene ≥97.0% (by GC)
4-Bromobenz[a]anthracene
Formula: C₁₈H₁₁Br
MW: 307.19 g/mol
MDL Number: MFCD25371993
CAS Number: 61921-39-9

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 215.0 to 220.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR