4,5-Bis(hydroxymethyl)-2-phenylimidazole ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

B4932-1G B4932-5G
TCB4932-1GEA 187 USD
TCB4932-1G TCB4932-5G
4,5-Bis(hydroxymethyl)-2-phenylimidazole ≥95.0% (by HPLC, titration analysis)
4,5-Bis(hydroxymethyl)-2-phenylimidazole
Formula: C₁₁H₁₂N₂O₂
MDL Number: MFCD00047017
CAS Number: 61698-32-6
EINECS: 262-911-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR