2-Hydroxybenzaldehyde Phenylhydrazone ≥98.0% (by GC)
Supplier: TCI America
H1478-25G
H1478-5G
TCH1478-25GEA
213
USD
TCH1478-25G
TCH1478-5G
2-Hydroxybenzaldehyde Phenylhydrazone ≥98.0% (by GC)
2-Hydroxybenzaldehyde Phenylhydrazone
Formula:
C₁₃H₁₂N₂O Boiling Pt: 234°C |
MDL Number:
MFCD00136159 CAS Number: 614-65-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 142.0 to 146.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...