Tetrakis(4-bromophenyl)ethylene ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
1,1,2,2-Tetrakis(4-bromophenyl)ethene
T3423-1G
T3423-5G
TCT3423-1GEA
128
USD
TCT3423-1G
TCT3423-5G
Tetrakis(4-bromophenyl)ethylene ≥97.0% (by GC)
Tetrakis(4-bromophenyl)ethylene
Formula:
C₂₆H₁₆Br₄ MW: 648.03 g/mol |
MDL Number:
MFCD00667761 CAS Number: 61326-44-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...