Verbascoside ≥97.0% (by HPLC)

Supplier: TCI America

V0153-10MG V0153-50MG
TCV0153-10MGEA 143.48 USD
TCV0153-10MG TCV0153-50MG
Verbascoside ≥97.0% (by HPLC)
Verbascoside
Formula: C₂₉H₃₆O₁₅
Storage Temperature: Refrigerator
MDL Number: MFCD00221751
CAS Number: 61276-17-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b92ba146d0c4c856d765c6848e53310c25b53606&vid=b92ba146d0c4c856d765c6848e53310c25b53606&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystaline
Purity (HPLC) min. 97.0 area%
Specific rotation [a]20/D -87.0 to -97.0 deg (C=0.1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR