4-(3,5-Dibromophenyl)-2,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

D4895-1G D4895-200MG
TCD4895-1GEA 103.6 USD
TCD4895-1G TCD4895-200MG
4-(3,5-Dibromophenyl)-2,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)
4-(3,5-Dibromophenyl)-2,6-diphenylpyrimidine
Formula: C₂₂H₁₄Br₂N₂
MDL Number: MFCD16619323
CAS Number: 607740-08-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 197.0 to 201.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR