9,3':6',9''-Tercarbazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

T3344-1G T3344-200MG
TCT3344-1GEA 212 USD
TCT3344-1G TCT3344-200MG
9,3':6',9''-Tercarbazole ≥98.0% (by HPLC, total nitrogen)
9,3':6',9''-Tercarbazole
Formula: C₃₆H₂₃N₃
Melting Pt: 285°C
MDL Number: MFCD29472452
CAS Number: 606129-90-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR