tert-Butylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
|
Warning
|
B5187-1G
B5187-5G
TCB5187-1GEA
29
USD
TCB5187-1G
TCB5187-5G
tert-Butylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
tert-Butylamine hydrobromide
Formula:
C₄H₁₁N·HBr |
MDL Number:
MFCD00674188 CAS Number: 60469-70-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...