5-Chloro-1-methylisatin ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 5-Chloro-1-methylindole-2,3-dione, 5-Chloro-1-methylindoline-2,3-dione, 5-Chloro-1-methyl-2,3-indolinedione

C3195-1G C3195-200MG
TCC3195-1GEA 403.8 USD
TCC3195-1G TCC3195-200MG
5-Chloro-1-methylisatin ≥98.0% (by GC)
5-Chloro-1-methylisatin
Formula: C₉H₆ClNO₂
MDL Number: MFCD02956479
CAS Number: 60434-13-1
EINECS: 262-231-5

Order Now

Specification Test Results

Appearance Orange to Red powder to crystal
Purity (GC) min. 98.0 %
Melting point 175.0 to 179.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR