9-(4-Bromophenyl)-3,6-di-tert-butylcarbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B5313-1G B5313-5G
TCB5313-1GEA 152 USD
TCB5313-1G TCB5313-5G
9-(4-Bromophenyl)-3,6-di-tert-butylcarbazole ≥98.0% (by GC)
9-(4-Bromophenyl)-3,6-di-tert-butylcarbazole
Formula: C₂₆H₂₈BrN
MDL Number: MFCD11617971
CAS Number: 601454-33-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 154.0 to 158.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR