Piperazine dihydrobromide ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Piperazinium Dibromide, Piperazinium bromide
P2490-1G
P2490-5G
TCP2490-1GEA
47
USD
TCP2490-1G
TCP2490-5G
Piperazine dihydrobromide ≥98.0% (by total nitrogen and titration analysis)
Piperazine dihydrobromide
Formula:
C₄H₁₀N₂·₂HBr Melting Pt: 360°C |
CAS Number:
59813-05-7 EINECS: 261-937-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...