Biotin-PEG2-NHS

Supplier: TCI America

Synonyms: N-Succinimidyl 9-(Biotinamido)-4,7-dioxanonanoate, 2,5-Dioxopyrrolidin-1-yl 3-(2-(2-(5-(2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)ethoxy)ethoxy)propanoate, Biotin-PEG2-NHS ester, 9-(Biotinamido)-4,7-dioxanonanoic acid N-Succinimidyl ester

S0955-100MG S0955-25MG
TCS0955-100MGEA 468 USD
TCS0955-100MG TCS0955-25MG
Biotin-PEG2-NHS
Biotin-PEG2-NHS
Formula: C₂₁H₃₂N₄O₈S
MW: 500.57 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00467154
CAS Number: 596820-83-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Elemental analysis (Carbon) 47.90 to 52.90 %
Elemental analysis (Nitrogen) 10.60 to 11.80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR