Pentabromobenzyl acrylate ≥98.0% (by GC)

Supplier: TCI America

P1877-1G P1877-5G
TCP1877-1GEA 86 USD
TCP1877-1G TCP1877-5G
Pentabromobenzyl acrylate ≥98.0% (by GC)
Pentabromobenzyl acrylate
Formula: C₁₀H₅Br₅O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00804467
CAS Number: 59447-55-1
EINECS: 261-767-7

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR