(Chloromethyl)trimethoxysilane ≥98.0% (by GC)

Supplier: TCI America
Danger

C3073-25G C3073-5G
TCC3073-25GEA 180.21 USD
TCC3073-25G TCC3073-5G
(Chloromethyl)trimethoxysilane ≥98.0% (by GC)
(Chloromethyl)trimethoxysilane
Formula: C₄H₁₁ClO₃Si
MW: 170.66 g/mol
Boiling Pt: 156°C
Flash Pt: -3°C
MDL Number: MFCD00468107
CAS Number: 5926-26-1
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR