Diphenhydramine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-(Benzhydryloxy)-N,N-dimethylethylamine, 2-(Benzhydryloxy)-N,N-dimethylethanamine, 2-Diphenylmethoxy-N,N-dimethylethylamine

D4744-25G D4744-5G
TCD4744-25GEA 134.26 USD
TCD4744-25G TCD4744-5G
Diphenhydramine ≥98.0% (by GC, titration analysis)
Diphenhydramine
Formula: C₁₇H₂₁NO
MW: 255.36 g/mol
MDL Number: MFCD00274173
CAS Number: 58-73-1
EINECS: 200-396-7
Merck Index: 14,03309

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR