2,4,6-Triphenylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

T3394-1G T3394-200MG
76220-608EA 226 USD
76220-608 76220-610
2,4,6-Triphenylpyridine ≥98.0% (by GC)
2,4,6-Triphenylpyridine
Formula: C₂₃H₁₇N
MDL Number: MFCD00014630
CAS Number: 580-35-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 139.0 to 143.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR