2',5'-Dimethoxypropiophenone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(2,5-Dimethoxyphenyl)-1-propanone

D4817-1G D4817-200MG
76219-708EA 188 USD
76219-708 76219-710
2',5'-Dimethoxypropiophenone ≥98.0% (by GC)
2',5'-Dimethoxypropiophenone
Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 168°C
MDL Number: MFCD00027013
CAS Number: 5803-30-5
EINECS: 227-357-7

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR