9-Acetylcarbazole ≥98.0% (by GC)

Supplier: TCI America

A3373-1G A3373-5G
TCA3373-1GEA 49 USD
TCA3373-1G TCA3373-5G
9-Acetylcarbazole ≥98.0% (by GC)
9-Acetylcarbazole
Formula: C₁₄H₁₁NO
MDL Number: MFCD00087027
CAS Number: 574-39-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR