9-[4-(tert-Butyl)phenyl]-9H-carbazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5620-1G
B5620-5G
TCB5620-1GEA
133
USD
TCB5620-1G
TCB5620-5G
9-[4-(tert-Butyl)phenyl]-9H-carbazole ≥98.0% (by GC)
9-[4-(tert-Butyl)phenyl]-9H-carbazole
Formula:
C₂₂H₂₁N |
MDL Number:
MFCD31618114 CAS Number: 57103-13-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 189.0 to 193.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...