3,6-Dimethoxy-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

D4833-1G D4833-200MG
TCD4833-1GEA 247.9 USD
TCD4833-1G TCD4833-200MG
3,6-Dimethoxy-9H-carbazole ≥98.0% (by GC)
3,6-Dimethoxy-9H-carbazole
Formula: C₁₄H₁₃NO₂
MDL Number: MFCD00626046
CAS Number: 57103-01-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR