9,9'-Diphenyl-9H,9'H-3,3'-bicarbazole ≥98.0% (by HPLC)

Supplier: TCI America
Warning

D4903-1G D4903-5G
TCD4903-1GEA 259 USD
TCD4903-1G TCD4903-5G
9,9'-Diphenyl-9H,9'H-3,3'-bicarbazole ≥98.0% (by HPLC)
9,9'-Diphenyl-9H,9'H-3,3'-bicarbazole
Formula: C₃₆H₂₄N₂
MW: 484.60 g/mol
MDL Number: MFCD29905037
CAS Number: 57102-62-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 204.0 to 208.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR