N-(4-Fluorobenzylidene)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0891-1G F0891-5G
TCF0891-1GEA 57 USD
TCF0891-1G TCF0891-5G
N-(4-Fluorobenzylidene)aniline ≥98.0% (by GC, titration analysis)
N-(4-Fluorobenzylidene)aniline
Formula: C₁₃H₁₀FN
MW: 199.23 g/mol
Boiling Pt: 176°C
MDL Number: MFCD00017971
CAS Number: 5676-81-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 42.0 to 46.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR