4-Benzoylphenyl methacrylate ≥98.0% (by GC)

Supplier: TCI America

B6151-25G B6151-5G
TCB6151-25GEA 234 USD
TCB6151-25G TCB6151-5G
4-Benzoylphenyl methacrylate ≥98.0% (by GC)
4-Benzoylphenyl methacrylate
Formula: C₁₇H₁₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD30470334
CAS Number: 56467-43-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR