IR 676 iodide ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

I1030-1G I1030-200MG
TCI1030-1GEA 385 USD
TCI1030-1G TCI1030-200MG
IR 676 iodide ≥97.0% (by HPLC, total nitrogen)
IR 676 iodide
Formula: C₃₅H₃₅IN₂
MDL Number: MFCD07433812
CAS Number: 56289-64-6

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Lambda max. 675.0 to 685.0 nm (EtOH)
Absorbance (E1%1cm) min. 3400 (EtOH, 675.0 to 685.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR