2-Bromo-4,6-diphenylpyrimidine ≥97.0% (by GC)

Supplier: TCI America
Warning

B4662-1G B4662-200MG
TCB4662-1GEA 345.2 USD
TCB4662-1G TCB4662-200MG
2-Bromo-4,6-diphenylpyrimidine ≥97.0% (by GC)
2-Bromo-4,6-diphenylpyrimidine
Formula: C₁₆H₁₁BrN₂
MDL Number: MFCD00234884
CAS Number: 56181-49-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR