2,6-Dibromo-9,10-di(2-naphthyl)anthracene ≥98.0% (by HPLC)
Supplier: TCI America
D4646-1G
D4646-200MG
TCD4646-1GEA
372.4
USD
TCD4646-1G
TCD4646-200MG
2,6-Dibromo-9,10-di(2-naphthyl)anthracene ≥98.0% (by HPLC)
2,6-Dibromo-9,10-di(2-naphthyl)anthracene
Formula:
C₃₄H₂₀Br₂ |
MDL Number:
MFCD16660916 CAS Number: 561064-15-1 |
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...