Pyrrolidine hydrobromide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Pyrrolidinium bromide
P2484-1G
P2484-5G
TCP2484-1GEA
59
USD
TCP2484-1G
TCP2484-5G
Pyrrolidine hydrobromide ≥98.0% (by titrimetric analysis)
Pyrrolidine hydrobromide
Formula:
C₄H₉N·HBr MW: 152.04 g/mol |
CAS Number:
55810-80-5 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...