Arprinocid ≥95.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: Arpocox, 9-(2-Chloro-6-fluorobenzyl)-9H-purin-6-amine

A3146-250MG A3146-50MG
76219-310EA 411 USD
76219-310 76219-312
Arprinocid ≥95.0% (by HPLC)
Arprinocid
Formula: C₁₂H₉ClFN₅
MW: 277.69 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00867210
CAS Number: 55779-18-5
EINECS: 259-817-8

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 24.00 to 26.50 %
Melting point 242.0 to 247.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR