AMAS (N-Succinimidyl maleimidoacetate)

Supplier: TCI America

Synonyms: Maleimidoacetic acid N-hydroxysuccinimide ester, AMAS, N-(alpha-Maleimidoacetoxy)succinimide, Maleimidoacetic acid N-Succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl 2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)acetate

S0968-100MG S0968-20MG
TCS0968-100MGEA 315 USD
TCS0968-100MG TCS0968-20MG
AMAS (N-Succinimidyl maleimidoacetate)
AMAS (N-Succinimidyl maleimidoacetate)
Formula: C₁₀H₈N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00133523
CAS Number: 55750-61-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Elemental analysis (Nitrogen) 10.50 to 11.50 %
Melting point 186.0 to 191.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR