5,6-Diaminobenzimidazolinone ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5,6-Diamino-1H-benzo[d]imidazol-2(3H)-one, 5,6-Diamino-2-hydroxybenzimidazole

D4533-1G D4533-5G
TCD4533-1GEA 37 USD
TCD4533-1G TCD4533-5G
5,6-Diaminobenzimidazolinone ≥95.0% (by HPLC, titration analysis)
5,6-Diaminobenzimidazolinone
Formula: C₇H₈N₄O
MW: 164.17 g/mol
MDL Number: MFCD00994340
CAS Number: 55621-49-3
EINECS: 259-728-4

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR