2-(p-Tolyl)indole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-(4-Methylphenyl)indole, 2-(4-Methylphenyl)-1H-indole

T3351-1G T3351-5G
76220-576EA 173 USD
76220-576 76220-578
2-(p-Tolyl)indole ≥98.0% (by GC)
2-(p-Tolyl)indole
Formula: C₁₅H₁₃N
MW: 207.28 g/mol
MDL Number: MFCD00087262
CAS Number: 55577-25-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 220.0 to 224.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR