(4-Bromophenyl)diphenylphosphine oxide ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5459-1G
B5459-5G
76219-622EA
88
USD
76219-622
76219-624
(4-Bromophenyl)diphenylphosphine oxide ≥98.0% (by GC)
(4-Bromophenyl)diphenylphosphine oxide
Formula:
C₁₈H₁₄BrOP MW: 357.19 g/mol |
MDL Number:
MFCD00411450 CAS Number: 5525-40-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 153.0 to 157.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...