3-Aminobenzofuran-2-carboxamide ≥98.0% (by GC)

Supplier: TCI America
Warning

A2832-1G A2832-5G
TCA2832-1GEA 49 USD
TCA2832-1G TCA2832-5G
3-Aminobenzofuran-2-carboxamide ≥98.0% (by GC)
3-Aminobenzofuran-2-carboxamide
Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
MDL Number: MFCD00457147
CAS Number: 54802-10-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 157.0 to 161.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR